1-(tert-Butyl)-2-ethyl-3-bromo-1H-indole-1,2-dicarboxylate ≥95%
Supplier: Matrix Scientific
046285-500MG
046285-1G
101874-950EA
276.92
USD
101874-950
101874-952
1-(tert-Butyl)-2-ethyl-3-bromo-1H-indole-1,2-dicarboxylate ≥95%
1-(tert-Butyl)-2-ethyl-3-bromo-1H-indole-1,2-dicarboxylate
Hazard: Irritant
Formula:
C₁₆H₁₈BrNO₄ |
MDL Number:
MFCD12025851 CAS Number: 1135283-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...