5-Bromo-6-cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%

Supplier: Matrix Scientific

046267-500MG 046267-1G
101874-886EA 921.54 USD
101874-886 101874-888
5-Bromo-6-cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
5-Bromo-6-cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-amine

Hazard: Irritant

Formula: C₁₀H₁₁BrN₄
MDL Number: MFCD11840982
CAS Number: 1135283-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR