Methyl-1-methyl-4-propionyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

046254-500MG 046254-1G
101874-842EA 556.92 USD
101874-842 101874-844
Methyl-1-methyl-4-propionyl-1H-pyrrole-2-carboxylate ≥95%
Methyl-1-methyl-4-propionyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD11840973
CAS Number: 1135283-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR