(E)-3-(2-Morpholino-5-nitrophenyl)-2-propenoic acid ≥95%

Supplier: Matrix Scientific

046250-500MG 046250-1G
101874-826EA 278.46 USD
101874-826 101874-828
(E)-3-(2-Morpholino-5-nitrophenyl)-2-propenoic acid ≥95%
(E)-3-(2-Morpholino-5-nitrophenyl)-2-propenoic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₅
MDL Number: MFCD06205585
CAS Number: 300665-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR