(2-(1H-1,2,4-Triazol-1-yl)phenyl)methanol ≥95%

Supplier: Matrix Scientific

046238-500MG 046238-1G
101874-784EA 375.38 USD
101874-784 101874-786
(2-(1H-1,2,4-Triazol-1-yl)phenyl)methanol ≥95%
(2-(1H-1,2,4-Triazol-1-yl)phenyl)methanol

Hazard: Irritant

Formula: C₉H₉N₃O
MW: 175.1900 g/mol
MDL Number: MFCD05864820
CAS Number: 914349-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR