3-Oxo-3-(1H-pyrrol-2-yl)propanenitrile ≥95%

Supplier: Matrix Scientific

046200-500MG 046200-1G
101874-652EA 415.38 USD
101874-652 101874-654
3-Oxo-3-(1H-pyrrol-2-yl)propanenitrile ≥95%
3-Oxo-3-(1H-pyrrol-2-yl)propanenitrile

Hazard: Irritant

Formula: C₇H₆N₂O
MDL Number: MFCD07369265
CAS Number: 90908-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR