2-[4-(2-Hydroxyethyl)piperazino]-1-(2-thienyl)-1-propanone ≥95%

Supplier: Matrix Scientific

046199-500MG 046199-1G 046199-5G
101874-646EA 415.38 USD
101874-646 101874-648 101874-650
2-[4-(2-Hydroxyethyl)piperazino]-1-(2-thienyl)-1-propanone ≥95%
2-[4-(2-Hydroxyethyl)piperazino]-1-(2-thienyl)-1-propanone

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂S
MDL Number: MFCD02931014
CAS Number: 4506-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR