Ethyl-4-[1-methyl-2-oxo-2-(2-thienyl)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

046196-500MG 046196-1G 046196-5G
101874-632EA 375.38 USD
101874-632 101874-634 101874-636
Ethyl-4-[1-methyl-2-oxo-2-(2-thienyl)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Ethyl-4-[1-methyl-2-oxo-2-(2-thienyl)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₃S
MDL Number: MFCD09972217
CAS Number: 1017782-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR