6-Chloro-N-methyl-2-phenyl-4-pyrimidinamine ≥95%

Supplier: Matrix Scientific

046187-500MG 046187-1G 046187-5G
101874-588EA 375.38 USD
101874-588 101874-590 101874-592
6-Chloro-N-methyl-2-phenyl-4-pyrimidinamine ≥95%
6-Chloro-N-methyl-2-phenyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃
MW: 219.68 g/mol
MDL Number: MFCD00697490
CAS Number: 1017782-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR