Methyl-5-amino-2-fluorobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-amino-2-fluorobenzoate

046123-1G 046123-5G
101874-268EA 58.46 USD
101874-268 101874-270
Methyl-5-amino-2-fluorobenzenecarboxylate ≥95%
Methyl-5-amino-2-fluorobenzenecarboxylate

Hazard: Irritant

Formula: C₈H₈FNO₂
MW: 169.16 g/mol
MDL Number: MFCD09037918
CAS Number: 56741-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR