Methyl-4-propionyl-1H-pyrrole-2-carboxylate ≥97%

Supplier: Matrix Scientific

046110-500MG 046110-1G
101874-214EA 556.92 USD
101874-214 101874-216
Methyl-4-propionyl-1H-pyrrole-2-carboxylate ≥97%
Methyl-4-propionyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD11840964
CAS Number: 1135282-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR