4-Chloro-1-benzothiophene-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

046099-500MG 046099-1G
101874-162EA 307.69 USD
101874-162 101874-164
4-Chloro-1-benzothiophene-2-carbaldehyde ≥95%
4-Chloro-1-benzothiophene-2-carbaldehyde

Hazard: Irritant

Formula: C₉H₅ClOS
MDL Number: MFCD11840956
CAS Number: 216575-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR