3-(tert-Butyl)-5-(chloromethyl)-1,2,4-oxadiazole ≥95%
Supplier: Matrix Scientific
046098-500MG
046098-1G
101874-158EA
375.38
USD
101874-158
101874-160
3-(tert-Butyl)-5-(chloromethyl)-1,2,4-oxadiazole ≥95%
3-(tert-Butyl)-5-(chloromethyl)-1,2,4-oxadiazole
Hazard: Irritant
Formula:
C₇H₁₁ClN₂O MW: 174.63 g/mol |
MDL Number:
MFCD09738869 CAS Number: 944901-64-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...