Methyl-6-formylnicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-formylnicotinate

046081-500MG 046081-1G
101874-072EA 138.46 USD
101874-072 101874-074
Methyl-6-formylnicotinate ≥95%
Methyl-6-formylnicotinate

Hazard: Irritant

Formula: C₈H₇NO₃
MW: 165.1500 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD08275010
CAS Number: 10165-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR