Ethyl-1-(4-formylphenyl)-4-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
046047-500MG
046047-1G
046047-5G
101873-896EA
140
USD
101873-896
101873-898
101873-900
Ethyl-1-(4-formylphenyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(4-formylphenyl)-4-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₉NO₃ |
MDL Number:
MFCD05864672 CAS Number: 85345-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...