2-Morpholinobenzaldehyde ≥95%

Supplier: Matrix Scientific

046044-500MG 046044-1G 046044-5G
101873-884EA 278.46 USD
101873-884 101873-886 101873-888
2-Morpholinobenzaldehyde ≥95%
2-Morpholinobenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MW: 191.2300 g/mol
MDL Number: MFCD00662562
CAS Number: 58028-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR