5-(2-Bromophenyl)oxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(2-Bromophenyl)-1,3-oxazole

045998-500MG 045998-1G
101873-654EA 126.15 USD
101873-654 101873-656
5-(2-Bromophenyl)oxazole ≥95%
5-(2-Bromophenyl)oxazole

Hazard: Irritant

Formula: C₉H₆BrNO
MW: 224.0540 g/mol
MDL Number: MFCD05668985
CAS Number: 328270-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR