Methyl-5-amino-2-(4-benzylpiperazino)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

045989-500MG 045989-1G 045989-5G
101873-620EA 556.92 USD
101873-620 101873-622 101873-624
Methyl-5-amino-2-(4-benzylpiperazino)benzenecarboxylate ≥95%
Methyl-5-amino-2-(4-benzylpiperazino)benzenecarboxylate

Hazard: Irritant

Formula: C₁₉H₂₃N₃O₂
MDL Number: MFCD03844862
CAS Number: 886360-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR