1-(4-Benzylpiperazino)-1-propanone ≥95%

Supplier: Matrix Scientific

045967-500MG 045967-1G 045967-5G
101873-506EA 181.54 USD
101873-506 101873-508 101873-510
1-(4-Benzylpiperazino)-1-propanone ≥95%
1-(4-Benzylpiperazino)-1-propanone

Hazard: Irritant

Formula: C₁₄H₂₀N₂O
MW: 232.33 g/mol
MDL Number: MFCD03381901
CAS Number: 314728-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR