1-{2-[2-(Trifluoromethyl)anilino]-1,3-thiazol-5-yl}-1-ethanone ≥95%

Supplier: Matrix Scientific

045923-500MG 045923-1G
101873-280EA 747.69 USD
101873-280 101873-282
1-{2-[2-(Trifluoromethyl)anilino]-1,3-thiazol-5-yl}-1-ethanone ≥95%
1-{2-[2-(Trifluoromethyl)anilino]-1,3-thiazol-5-yl}-1-ethanone

Hazard: Irritant

Formula: C₁₂H₉F₃N₂OS
MDL Number: MFCD11840951
CAS Number: 1135282-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR