Ethyl-2-cyano-3-(4-nitrophenyl)acrylate ≥95%

Supplier: Matrix Scientific

045899-500MG 045899-1G
101873-168EA 181.54 USD
101873-168 101873-170
Ethyl-2-cyano-3-(4-nitrophenyl)acrylate ≥95%
Ethyl-2-cyano-3-(4-nitrophenyl)acrylate

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₄
MW: 246.22 g/mol
MDL Number: MFCD00024795
CAS Number: 2017-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR