4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl [4,4'-bipiperidine]-1-carboxylate, 4,4'-Bipiperidine, 1-BOC protected, N-Boc-4,4'-bipiperidine

045893-500MG 045893-1G
101873-142EA 195.38 USD
101873-142 101873-144
4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine ≥95%
4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine

Hazard: Irritant

Formula: C₁₅H₂₈N₂O₂
MW: 268.4 g/mol
MDL Number: MFCD02179169
CAS Number: 171049-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR