Methyl-2-(2-butyl-1,3-benzoxazol-5-yl)acetate ≥95%

Supplier: Matrix Scientific

045768-500MG 045768-1G
101872-492EA 747.69 USD
101872-492 101872-494
Methyl-2-(2-butyl-1,3-benzoxazol-5-yl)acetate ≥95%
Methyl-2-(2-butyl-1,3-benzoxazol-5-yl)acetate

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MDL Number: MFCD05663830
CAS Number: 886361-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR