1-(6-Fluoro-2-pyridinyl)tetrahydro-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(6-Fluoropyridin-2-yl)piperidin-4-one

045580-500MG 045580-1G 045580-5G
101871-586EA 278.46 USD
101871-586 101871-588 101871-590
1-(6-Fluoro-2-pyridinyl)tetrahydro-4(1H)-pyridinone ≥95%
1-(6-Fluoro-2-pyridinyl)tetrahydro-4(1H)-pyridinone

Hazard: Irritant

Formula: C₁₀H₁₁FN₂O
MW: 194.21 g/mol
MDL Number: MFCD00665213
CAS Number: 339096-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR