3-Ethoxy-2-(methylsulfonyl)acrylonitrile ≥95%

Supplier: Matrix Scientific

044235-500MG 044235-1G
101864-864EA 346.15 USD
101864-864 101864-866
3-Ethoxy-2-(methylsulfonyl)acrylonitrile ≥95%
3-Ethoxy-2-(methylsulfonyl)acrylonitrile

Hazard: Irritant

Formula: C₆H₉NO₃S
MDL Number: MFCD11553018
CAS Number: 104007-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR