2-{[4-(Dimethylamino)phenyl]methylene}-1-hydrazinecarbothioamide ≥95%

Supplier: Matrix Scientific

044163-500MG 044163-1G
101864-476EA 278.46 USD
101864-476 101864-478
2-{[4-(Dimethylamino)phenyl]methylene}-1-hydrazinecarbothioamide ≥95%
2-{[4-(Dimethylamino)phenyl]methylene}-1-hydrazinecarbothioamide

Hazard: Irritant

Formula: C₁₀H₁₄N₄S
MDL Number: MFCD00276108
CAS Number: 2929-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR