1-(3-Nitro-2-pyridinyl)-4-piperidinol ≥95%

Supplier: Matrix Scientific

044118-500MG 044118-1G 044118-5G
101864-238EA 278.46 USD
101864-238 101864-240 101864-242
1-(3-Nitro-2-pyridinyl)-4-piperidinol ≥95%
1-(3-Nitro-2-pyridinyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₃
MDL Number: MFCD00974220
CAS Number: 338411-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR