1-(2-Bromophenyl)pyrrole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2-Bromophenyl)-1H-pyrrole

043993-500MG 043993-1G 043993-5G
101863-570EA 181.54 USD
101863-570 101863-572 101863-574
1-(2-Bromophenyl)pyrrole ≥97%
1-(2-Bromophenyl)pyrrole

Hazard: Irritant

Formula: C₁₀H₈BrN
MW: 222.0800 g/mol
MDL Number: MFCD02665226
CAS Number: 69907-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR