Ethyl-2-(4-benzylpiperazino)-2-phenylacetate ≥95%
Supplier: Matrix Scientific
043973-500MG
043973-1G
043973-5G
101863-470EA
278.46
USD
101863-470
101863-472
101863-474
Ethyl-2-(4-benzylpiperazino)-2-phenylacetate ≥95%
Ethyl-2-(4-benzylpiperazino)-2-phenylacetate
Hazard: Irritant
Formula:
C₂₁H₂₆N₂O₂ |
MDL Number:
MFCD04117802 CAS Number: 863305-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...