1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Acetyl-5-chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole

043963-500MG 043963-1G
101863-430EA 747.69 USD
101863-430 101863-432
1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanone ≥95%
1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanone

Hazard: Irritant

Formula: C₇H₆ClF₃N₂O
MW: 226.59 g/mol
MDL Number: MFCD04117795
CAS Number: 863305-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR