(4-Bromophenyl)(cyclopropyl)ketone ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Bromophenyl)(cyclopropyl)methanone, (4-Bromophenyl)cyclopropylmethanone, (4-Bromobenzoyl)cyclopropane, 4-Bromophenyl Cyclopropyl ketone

043959-1G 043959-5G
101863-414EA 29.23 USD
101863-414 101863-416
(4-Bromophenyl)(cyclopropyl)ketone ≥95%
(4-Bromophenyl)(cyclopropyl)ketone

Hazard: Irritant

Formula: C₁₀H₉BrO
MW: 225.09 g/mol
MDL Number: MFCD00019227
CAS Number: 6952-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR