4-(4-Nitrophenyl)piperidine ≥95%

Supplier: Matrix Scientific

043957-500MG 043957-1G 043957-5G
101863-402EA 267.69 USD
101863-402 101863-404 101863-406
4-(4-Nitrophenyl)piperidine ≥95%
4-(4-Nitrophenyl)piperidine

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD06804549
CAS Number: 26905-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR