Methyl-2-(4-hydroxyphenoxy)acetate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-(4-hydroxyphenoxy)acetate
043897-500MG
043897-1G
101863-106EA
846.15
USD
101863-106
101863-108
Methyl-2-(4-hydroxyphenoxy)acetate ≥95%
Methyl-2-(4-hydroxyphenoxy)acetate
Hazard: Irritant
Formula:
C₉H₁₀O₄ MW: 182.18 g/mol |
MDL Number:
MFCD11046458 CAS Number: 70067-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...