1,3-Dimethyl-5-phenoxy-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

043890-500MG 043890-1G
101863-086EA 921.54 USD
101863-086 101863-088
1,3-Dimethyl-5-phenoxy-1H-pyrazole-4-carbaldehyde ≥95%
1,3-Dimethyl-5-phenoxy-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD07698583
CAS Number: 109925-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR