(2-phenoxyphenyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Phenoxyphenyl)methanol, 2-Phenoxybenzyl alcohol

043880-500MG 043880-1G
101863-036EA 186.15 USD
101863-036 101863-038
(2-phenoxyphenyl)methanol ≥95%
(2-phenoxyphenyl)methanol

Hazard: Irritant

Formula: C₁₃H₁₂O₂
MW: 200.237 g/mol
MDL Number: MFCD00017297
CAS Number: 13807-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR