1-Cyclopropyl-4,4,4-trifluoro-1,3-butanedione ≥95%

Supplier: Matrix Scientific

043871-500MG 043871-1G
101862-992EA 375.38 USD
101862-992 101862-994
1-Cyclopropyl-4,4,4-trifluoro-1,3-butanedione ≥95%
1-Cyclopropyl-4,4,4-trifluoro-1,3-butanedione

Hazard: Irritant

Formula: C₇H₇F₃O₂
MDL Number: MFCD03419771
CAS Number: 30923-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR