Ethyl-2-[(benzyloxy)imino]-3-oxobutanoate ≥95%

Supplier: Matrix Scientific

043816-500MG 043816-1G 043816-5G
101862-708EA 278.46 USD
101862-708 101862-710 101862-712
Ethyl-2-[(benzyloxy)imino]-3-oxobutanoate ≥95%
Ethyl-2-[(benzyloxy)imino]-3-oxobutanoate

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD08056614
CAS Number: 289882-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR