4-(2-Oxo-2,3-dihydro-1H-1,3-benzimidazol-1-yl)butanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-Oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)butanoic acid

043766-1G 043766-5G
101862-470EA 15.38 USD
101862-470 102833-124
4-(2-Oxo-2,3-dihydro-1H-1,3-benzimidazol-1-yl)butanoic acid ≥95%
4-(2-Oxo-2,3-dihydro-1H-1,3-benzimidazol-1-yl)butanoic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD06656423
CAS Number: 3273-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR