Ethyl-1-(3-formyl-2-pyridinyl)-4-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
043742-1G
043742-5G
101862-348EA
698.46
USD
101862-348
101862-350
Ethyl-1-(3-formyl-2-pyridinyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(3-formyl-2-pyridinyl)-4-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₈N₂O₃ MW: 262.31 g/mol |
MDL Number:
MFCD06200933 CAS Number: 886361-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...