Ethyl-1-(1,1-dimethyl-2-oxoethyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

043710-500MG 043710-1G 043710-5G
101862-176EA 556.92 USD
101862-176 101862-178 101862-180
Ethyl-1-(1,1-dimethyl-2-oxoethyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(1,1-dimethyl-2-oxoethyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₂H₂₁NO₃
MW: 227.31 g/mol
MDL Number: MFCD05663786
CAS Number: 886360-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR