Ethyl-4-(4-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(4-aminophenyl)piperazine-1-carboxylate

043705-500MG 043705-1G
101862-148EA 556.92 USD
101862-148 101862-150
Ethyl-4-(4-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Ethyl-4-(4-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₂
MW: 249.32 g/mol
MDL Number: MFCD11848963
CAS Number: 16154-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR