Methyl-1-(1-benzyl-4-piperidinyl)-5-oxo-3-pyrrolidinecarboxylate ≥95%

Supplier: Matrix Scientific

043670-500MG 043670-1G 043670-5G
101861-988EA 181.54 USD
101861-988 101861-990 101861-992
Methyl-1-(1-benzyl-4-piperidinyl)-5-oxo-3-pyrrolidinecarboxylate ≥95%
Methyl-1-(1-benzyl-4-piperidinyl)-5-oxo-3-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₃
MW: 316.4 g/mol
MDL Number: MFCD08689668
CAS Number: 937601-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR