5-Cyano-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(1-Methyl-1,2,3,6-tetrahydropyridin-4-yl)-1H-indole-5-carbonitrile

043648-500MG 043648-1G
101861-862EA 375.38 USD
101861-862 101861-864
5-Cyano-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole ≥95%
5-Cyano-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole

Hazard: Irritant

Formula: C₁₅H₁₅N₃
MW: 237.306 g/mol
MDL Number: MFCD08276951
CAS Number: 116480-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR