5-Methoxy-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole ≥95%

Supplier: Matrix Scientific

043647-500MG 043647-1G
101861-858EA 375.38 USD
101861-858 101861-860
5-Methoxy-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole ≥95%
5-Methoxy-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole

Hazard: Irritant

Formula: C₁₅H₁₈N₂O
MW: 242.32 g/mol
MDL Number: MFCD08276947
CAS Number: 55556-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR