2-[Benzyl(2-hydroxyethyl)amino]-1-ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2-(Benzylimino)diethanol

043644-1G 043644-5G 043644-25G
101861-846EA 47.69 USD
101861-846 101861-848 101861-850
2-[Benzyl(2-hydroxyethyl)amino]-1-ethanol ≥95%
2-[Benzyl(2-hydroxyethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₇NO₂
MW: 195.26 g/mol
MDL Number: MFCD00020587
CAS Number: 101-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR