2,4-Difluorophenyl propionate ≥95%

Supplier: Matrix Scientific

043563-500MG 043563-1G 043563-5G
101861-454EA 181.54 USD
101861-454 101861-456 101861-458
2,4-Difluorophenyl propionate ≥95%
2,4-Difluorophenyl propionate

Hazard: Irritant

Formula: C₉H₈F₂O₂
MDL Number: MFCD08443940
CAS Number: 924868-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR