Pentafluorophenyl 2-propanesulfonate ≥95%

Supplier: Matrix Scientific

043343-500MG 043343-1G 043343-5G
101860-346EA 375.38 USD
101860-346 101860-348 101860-350
Pentafluorophenyl 2-propanesulfonate ≥95%
Pentafluorophenyl 2-propanesulphonate

Hazard: Irritant

Formula: C₉H₇F₅O₃S
MDL Number: MFCD05975149
CAS Number: 886361-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR