[3-(Benzyloxy)-4-methoxyphenyl]methanol ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Benzyloxy-4-methoxybenzyl alcohol, 3-(Benzyloxy)-4-methoxybenzyl alcohol

043292-500MG 043292-1G
101860-080EA 278.46 USD
101860-080 101860-082
[3-(Benzyloxy)-4-methoxyphenyl]methanol ≥97%
[3-(Benzyloxy)-4-methoxyphenyl]methanol

Hazard: Irritant

Formula: C₁₅H₁₆O₃
MW: 244.29 g/mol
MDL Number: MFCD00183283
CAS Number: 1860-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR