3-(tert-Butoxy)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-tert-Butoxybenzoic acid
043275-500MG
043275-1G
101860-020EA
1216.92
USD
101860-020
101860-022
3-(tert-Butoxy)benzenecarboxylic acid ≥95%
3-(tert-Butoxy)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₄O₃ MW: 194.23 g/mol |
MDL Number:
MFCD03412397 CAS Number: 15360-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...