Ethyl-2-(1-methyl-2,5-dioxotetrahydro-4H-imidazol-4-yliden)acetate ≥95%

Supplier: Matrix Scientific

043225-500MG 043225-1G 043225-5G
101859-772EA 184.62 USD
101859-772 101859-774 101859-776
Ethyl-2-(1-methyl-2,5-dioxotetrahydro-4H-imidazol-4-yliden)acetate ≥95%
Ethyl-2-(1-methyl-2,5-dioxotetrahydro-4H-imidazol-4-yliden)acetate

Hazard: Irritant

Formula: C₈H₁₀N₂O₄
MDL Number: MFCD09607892
CAS Number: 712-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR